missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 20 Germination Test Paper

Satisfies the requirements for the determination of germination capability according to ISTA (International Seed Testing Association). Sartorius™ Grade 20 Germination Test Paper is wet-strengthened and the special structure prevents fine seed roots from growing through the paper.

Brand:  Sartorius™ FT-2003532000110

27009.54 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11770815

  • 27100.00 SEK / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for ensuring optimal moisture content for the most diverse types of seeds and germination forms
  • Mainly applied with corn, sugar beets, wheat, barley and various grasses
  • Can also be used for all other seed types
  • pH ranges between 6.0 and 7.5
  • 112 rods with 9 pleated strips with 50 double pleats of 20mm depth
TRUSTED_SUSTAINABILITY
Specifications

Specifications

115 g/m2
White, non-sterile, wet-strengthened
Cotton Linters, Refined Pulp
20
0.26 mm
2000 x 110 mm
White
Pleated
Determination of germination capability
1000 Pk.
Germination Test Paper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.