missing translation for 'onlineSavingsMsg'
Learn More

Munktell Filtrak™ Grade 1600 Pulp Testing Filter Paper

For physical testing in accordance with ISO Test Method 5269-1: 2005, Part 1

Brand:  Munktell Filtrak™ 143040

Additional Details : Weight : 0.97200kg

Product Code. 11337254

  • 1821.48 SEK / Pack of 200
In Stock
to see stock.
Add to basket

Description

Description

  • Basis Weight: 250±25g/m2
Specifications

Specifications

Disc
1600
170 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers