Learn More
Trajan Scientific and Medical™ Graphite Ferrules for Shimadzu
Provide leak-free system connections, leading to improved chromatography for non-MS systems.
Brand:  Trajan Scientific and Medical™ 0726080
Product Code. 11942627
  • 1965.00 SEK / Pack of 10
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Specifications
Specifications
Graphite Ferrule for Shimadzu 0.1-0.32mm ID Column
GC14 A, GC17 A, GC2010 and GC2014 Detector / Injectors
Graphite
0.10 to 0.32 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan Scientific and Medical™ Graphite Ferrules for Shimadzu > Graphite Ferrules for Shimadzu