Learn More
Thermo Scientific Chemicals Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
5390.00 SEK - 6020.00 SEK
Specifications
Description | Unpurified extruded graphite |
---|---|
Material | Graphite |
For Use With (Application) | Metallurgy |
Product Type Specs | Graphite fusion crucible |
Shape | Drillpoint |
Product Code | Brand | Capacity (Metric) | Shape | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Shape | Quantity | Price | Quantity & Availability | |||
15466345
|
Thermo Scientific Chemicals
040794.KD |
7.5 cc | Drillpoint | 50pc |
6020.00 SEK
Pack of 50 |
In Stock
Log in to see stock. |
|||
15426055
|
Thermo Scientific Chemicals
014747.UB |
7.88 cc | Drillpoint | 36pc |
5390.00 SEK
Pack of 36 |
In Stock
Log in to see stock. |
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Unpurified extruded graphite | |
Metallurgy | |
Drillpoint |
Graphite | |
Graphite fusion crucible |