Learn More
Eppendorf™ Grease
Grease pistons when pipette has become extremely dirty. Eppendorf™ Grease is available in 45g tubes, for use with a variety of pipettes.
Brand:  Eppendorf™ 0013019231
Additional Details : Weight : 0.04680kg
Product Code. 10124621
  • 1115.00 SEK / 45g
In Stock
to see stock.
Description
Description
  • Silicone grease for pipettes
  • Clean pipette with organic solvents; apply grease following cleaning
Specifications
Specifications
1 Each
45 g