Learn More
Keystone Cleanroom Products Green Cellulose Sterilization Wrap
GREENER_CHOICE
When secured with autoclave tape or elasticized bands, these blue GHD wrappers provide an effective microbial barrier from external sources of contamination.
Brand:  Keystone Cleanroom Products GHDSHEET0303
 View more versions of this product
Product Code. 30089993
  • 2165.00 SEK / Pack of 3000
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Designed for steam sterilization; but also compatible with ETO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Superior bacterial barrier properties compared to linen
  • Sterilization tapes adhere firmly and remove easily
  • Ideal choice for light trays, small basins and towels
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO certified cleanrooms
Specifications
Specifications
57 gsm
Autoclavable
None
Single-use
Cellulose
Yes
121°C to 135°C
7.6 cm
7.6 cm
Sterilization Wrap (Heavy-duty)
CoC
Green
Steam, Gamma, or EtO Sterilization
Non-sterile
3000/Cs.
3 in.
3 in.