Learn More
GSG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565074
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: QHRRRFFNSSG SSDASIGDPS QSDDPDDPDD PDFPGSPVRR RRRCPGGRVP KDRPSLTVTP KRWKLRARPS LTVTPRRLGL RARPPQKCST P Highest antigen sequence identity to the following orthologs - mouse 38%, rat 41%.
GSG2 (haspin) is a conserved kinase required for mitosis. GSG2 phosphorylates histone H3 and is viewed as a potential therapeutic target for oncology. GSG2 is required for normal alignment of chromosomes at metaphase.Specifications
GSG2 | |
Polyclonal | |
Unconjugated | |
HASPIN | |
germ cell associated 2 (haspin); germ cell associated 2, haspin; germ cell-specific gene 2 protein; GSG2; haploid germ cell-specific nuclear protein kinase; HASPIN; H-haspin; Serine/threonine-protein kinase haspin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83903 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TF76 | |
HASPIN | |
Recombinant Human GSG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only