missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | GSPT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18322854
|
Bio-Techne
NBP3-04706-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18370724
|
Bio-Techne
NBP3-04706-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSPT1 Polyclonal specifically detects GSPT1 in Human, Rat samples. It is validated for Western Blot.Specifications
GSPT1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Rat | |
ERF3A, eRF3aFLJ38048, ETF3A, eukaryotic peptide chain release factor GTP-binding subunit ERF3A, Eukaryotic peptide chain release factor subunit 3a, FLJ39067, G1 to S phase transition 1,551G9.2, G1 to S phase transition protein 1 homolog, GST1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 138-420 of human GSPT1 (NP_001123478.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS with 50% glycerol, pH7.3. | |
2935 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title