missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543186
This item is not returnable.
View return policy
Description
Peptide sequence: TWRQRVEAAV GEDLFQEAHE VILKAKDFPP ADPTIKQKLM PWVLAMIR Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 86%
GSTT1 Polyclonal antibody specifically detects GSTT1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GSTT1 | |
Polyclonal | |
Unconjugated | |
Gstt1 | |
AI255817; Glutathione S-transferase 1 (theta); glutathione S-transferase 5; glutathione S-transferase theta 1; glutathione S-transferase theta-1; glutathione S-transferase, theta 1; glutathione transferase T1-1; GST 5-5; GST class-theta-1; GSTT1; Gstt1-1; GSTYRS; similar to Glutathione S-transferase theta 1 (GST class-theta 1) (Glutathione transferase T1-1) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2952 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P30711 | |
Gstt1 | |
Synthetic peptide directed towards the C-terminal of human GSTT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |