missing translation for 'onlineSavingsMsg'
Learn More
Guanosine, 98%, Thermo Scientific Chemicals Product Code.: 11423734

Guanosine, 98%, Thermo Scientific Chemicals

Product Code. 11423734
50 g
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 11423734

Brand: Thermo Scientific Alfa Aesar A11328.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.

Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-00-3
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00010182
InChI Key NYHBQMYGNKIUIF-UUOKFMHZSA-N
Synonym guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
PubChem CID 6802
ChEBI CHEBI:16750
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point ∼250°C (decomposition)
Quantity 50 g
Beilstein 625911
Merck Index 14,4566
Solubility Information Soluble in Formic Acid:Water (1:1): 50mg/mL. Insoluble in methanol,diethyl ether.
Optical Rotation −72° (c=1.5 in 0.1N NaOH)
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 283.24
Percent Purity ≥98%
Chemical Name or Material Guanosine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243901 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog NumberA11328.18,A11328.14,A11328.22,A11328.30
Lot Number10238001 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11328.18,A11328.14,A11328.22,A11328.30
Lot Number10099306 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11328.18,A11328.14,A11328.22,A11328.30
Lot Number10215187 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11328.18,A11328.14,A11328.22,A11328.30
Lot Number10165186 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11328.18,A11328.14,A11328.22,A11328.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Guanosine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.