Learn More
MedChemExpress H-Gly-Gly-Pro-OH
MedChemExpress H-Gly-Gly-Pro-OH is a peptide with 3 amino acid.
Brand:  MedChemExpress HY-111922
Encompass_Preferred
Product Code. 16483116
  • 1758.29 SEK / 25mg
Explore more special offers
Specifications
97.0%
Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Peptides
Room temperature
14379-76-1
229.23 g/mol
25 mg
Glycyl-glycyl-L-proline
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.