missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) isopropoxide isopropanol adduct, 99%, Thermo Scientific Chemicals

Product Code. 15448907
Click to view available options
Quantity:
25 g
100 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2171-99-5
Molecular Formula C12H32HfO4
Molecular Weight (g/mol) 418.874
MDL Number MFCD00070460
InChI Key HMKGKDSPHSNMTM-UHFFFAOYSA-N
Synonym propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci
PubChem CID 74764872
IUPAC Name hafnium;propan-2-ol
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf]

Specifications

Linear Formula Hf(OOCH(CH3)2)4·HOCH(CH3)2
Quantity 25 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 474.94
Percent Purity 99%
Chemical Name or Material Hafnium(IV) isopropoxide isopropanol adduct

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.