Learn More
Hamilton™ Neuros Replacement Needles for Microliter/Gastight™ Syringes
Removable needles can be used with select Hamilton Removable Needle (RN) 700 and 1700 Series Neuros syringes
Brand:  Hamilton™ 65461-01
Additional Details : Weight : 0.04500kg
Product Code. 15162654
  • 2120.00 SEK / Each
Estimated Shipment: 13-08-2024
to see stock.
Description
Description
  • Special blunt needle point suited for neuroscience applications
  • 304 stainless steel
  • 33 gauge
  • Blunt point (Point Style 3)
  • Exposed needle length can be adjusted by the user from 0 to 20mm
  • Sold in 6-packs
Neuros replacement needles work with the following Hamilton 700 and 1700 Series syringes:
  • Model 75 RN Syringe (14-824-651)
  • Model 1701 RN Syringe (14-824-652)
  • Model 1702 RN Syringe (14-824-653)
  • Model 1705 RN Syringe (14-824-654)
  • Model 1710 RN Syringe (14-824-655)
Specifications
Specifications
Autoclavable
0.00015 in.
0.004 mm
Used for neuroscience applications and select Hamilton removable needle 700 and 1700 series 0.79μL neuros syringes
Removable Square/Blunt
33 G
0.0003 in.
0.008 mm
Replacement Needle