missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP™-3 Reversed Phase HPLC Columns

PRP-3 polymeric 300 Å columns for gradient separations of peptides and proteins

Brand:  Hamilton™ 79468

 View more versions of this product

Product Code. 11518084

  • 18290.00 SEK / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

PRP-3 polymeric reversed phase columns, packed with a poly(styrene-divinyl)benzene support, last longer than silica-based columns. Support is pressure stable up to 5,000 psi and crosslinked to prevent shrinkage or swelling when the mobile phase is changed. The support is entirely polymeric and there are no C18 groups to hydrolyze, column lifetime is longer.

Superior pH Stability

  • The inherent stability of PRP-3 columns allows their use for any protein purification method from pH 1 to 13. PRP-3 columns allow you to analyze protein purity at low and high pH extremes.
  • Protein Recovery
    • The 300Å pores on the PRP-3 support help ensure good recovery (>90%) of your protein. The highly inert poly(styrene-divinylbenzene) packing enhances protein recovery because there are no silanol groups on the support to cause irreversible protein adsorption
  • Wide Applicability
    • Easy to use and ideal for almost any protein separation, including pH conditions from 1 to 13, with organic modifier (0-100%) of virtually any type from routine to the unusual
  • Scaleup
    • A selection of column sizes are available from analytical to semi-prep and preparative scale. Sample scaleup is easy because the PRP-3 packing is consistent from analytical to preparative columns.
    • The short analytical (50mm) column is well suited for the gradient elution of high molecular weight proteins. The longer (150mm) column is well suited for smaller proteins.

    Product Features:

    • Particle size: 10μm and 12-20μL
    • Eight column internal diameters: 2.1 to 21.2μL
    • Two column materials: 316 stainless steel and PEEK
    • Analytical and semiprep/preparative guard columns
    • Custom hardware dimensions available

    Specifications

    Specifications

    Analytical Column
    305 mm
    pH 1 to 13, 0 to 100% aqueous, organic modifier
    1 to 13
    5°C to 85°C
    PRP-3
    Reversed Phase
    Stainless Steel
    7 mm
    5000 psi (345 bar)
    10 μm
    300 Å
    L21
    PSDVB (Polystyrene Divinylbenzene)
    Reversed Phase
    Column
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.