missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAPLN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558708
This item is not returnable.
View return policy
Description
Immunogen sequence: GLPFYNGFYY SNSANDQNLG NGHGKDLLNG VKLVVETPEE TLFTYQGASV ILPCRYRYEP ALVSPRRVRV KWWKLSEN Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
HAPLN3 is a 360 amino acid extracellular protein belonging to the HAPLN family and contains one Ig-like V-type (immunoglobulin-like) domain and two Link domains. This protein may function in hyaluronic acid binding and cell adhesion. It is either a secreted or an extracellular matrix protein involved in stabilizing the aggregates of proteoglycan monomers with hyaluronic acid in the extracellular cartilage matrix. HAPLN3 is widely expressed in most tissues with highest levels in spleen and placenta.Specifications
HAPLN3 | |
Polyclonal | |
Unconjugated | |
HAPLN3 | |
4930554N11Rik; EXLD1; extracellular link domain containing, 1; Hapln3; HsT19883; Hyaluronan and proteoglycan link protein 3; link protein 3; Lpr3; UNQ238/PRO271 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
145864 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96S86 | |
HAPLN3 | |
Recombinant protein corresponding to Human HAPLN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only