Learn More
New Pig Haz-Mat Pillows
Stop the spread of acids and bases.
Brand: New Pig PIL306
This item is not returnable.
View return policy
Description
- Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
- Excellent for quickly diking and stopping the spread of chemical spills
- Absorb and retain most acids, bases and unknown liquids
- Can be incinerated after use to reduce waste
Specifications
Pink | |
8.9 L | |
Hazmat Absorbent Pillow | |
8 L x 8 W x 1 in. H (20.3 x 20.3 x 2.54 cm) | |
8 in. | |
8 in. | |
1 in. | |
4 lb. |
30 oz. | |
Polypropylene | |
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives | |
8 x 8 x 1 in. (20.3 x 20.3 x 2.54 cm) | |
20.3 cm | |
20.3 cm | |
2.54 cm | |
1.8 kg |