Learn More
asecos™ Hazardous Substances Rack
The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.
Brand:  asecos™ 27869
 View more versions of this product
Product Code. 17816586
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Rack with galvanised shelf as storage levels
  • Collecting sump
  • 100 mm ground clearance for transport with pallet truck
Specifications
Specifications
Blue
205 L
Drum Rack
1236.00 mm
1236.00 mm
2100.00 mm
816.00 mm
102 kg
54.2 gal
Steel
48.66 in.
48.66 in.
82.68 in.
32.13 in.
224.87 lb.
Product Title
asecos™ Hazardous Substances Rack > Capacity: 205 L, 102 kg