missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 491025
This item is not returnable.
View return policy
Description
HDAC1 Polyclonal antibody specifically detects HDAC1 in Human, Mouse, Rat samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Western Blot.Specifications
HDAC1 | |
Polyclonal | |
Unconjugated | |
HDAC1 | |
DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3065, 433759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Western Blot | |
1.73 mg/mL | |
antibody with <0.1% sodium azide | |
Q13547, O09106 | |
HDAC1 | |
raised in rabbits against the C-terminal region of human HDAC1, using a KLH-conjugated synthetic peptide. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |