Learn More
VITLAB™ HDPE Measuring Scoop
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ HDPE Measuring Scoops are lighter than metal, don’t dent and feature good chemical resistance.
Brand: VITLAB™ 40393
Additional Details : Weight : 0.02600kg
Description
- Conical in shape with tapered filling edge
Specifications
Measuring Scoop | |
Conical | |
Natural |
HDPE | |
110 mL | |
Suitable as weighing |