Learn More
Kautex™ High Density Polyethylene Medium Mouth Bottles
Manufactured with HDPE for stability, strength and opacity. Kautex™ High Density Polyethylene Medium Mouth Bottles have an embossed graduated scale for permanent accurate measurement.
Brand:  Kautex™ 2000770073
 View more versions of this product
Product Code. 15961471
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Multi-purpose bottles for laboratories and industry
  • Embossed contents scale
  • Easy to fill and clean, leak-proof
  • PP screw closure
Application:
  • dispensing
  • sending samples
  • mailing
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 mL
39.2 mm
HDPE
1 Ea.
1 Ea.
Yes
203 mm
Polypropylene
97 mm
Natural
Dispensing, Sending Samples, Mailing, Transport and Storage
4/Cs.
Bottle
Yes
Round
Bottle
50 mm
Screw Cap