Learn More
Kautex™ HDPE Natural Rectangular Narrow Neck Bottle
Brand:  Kautex™ 2000070520
Additional Details : Weight : 0.97200kg
 View more versions of this product
Product Code. 15901391
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • High stability, ribbed sides for secure grip
  • Rectangular, natural color
  • Caps are not included; sold separately.
Specifications
Specifications
0.1 L
12.9 mm
HDPE
No
Square
Narrow Neck Bottle
Natural
Storage, Packaging, Dispensing, Mailing
Narrow
Narrow
110.5 mm