Learn More
Kautex™ HDPE Round Bottle
Use Kautex™ HDPE Round Bottle for many bottling needs, including cosmetic products, e.g. emulsions, shampoos, shower gels, creams and as a refill packaging. Order PP hinged closure separately;
Brand:  Kautex™ 2000073087
Additional Details : Weight : 2.88000kg
 View more versions of this product
Product Code. 10737732
  • 24.92 SEK / Pack of 72
Estimated Shipment: 27-08-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
  • Natural color
  • Stable and handy shape
  • PP hinged closure ordered separately
  • Hinged lid closure for single-handed use
Specifications
Specifications
1000 mL
Packaging, Storage and Transport
72Case
Bottle
White
White
Round
Polypropylene
89 mm
19.4 mm
HDPE
72/Pk.
1Each
Polypropylene
No
Bottle
22 mm
Hinged Lid