Learn More
JULABO™ Heat Exchanger Bath Cover
Bath cover with built-in heat exchanger
4845.00 SEK - 5705.00 SEK
Specifications
Description With built-in heat exchanger
Type Bath Cover
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15420622
View Documents
JULABO™
9970240
CORIO™ CD-BC4, CORIO™ CD-BC6, CORIO™ CP-200F, CORIO™ CP-201F, CORIO™ CP-300F, CORIO™ CP-BC4, CORIO™ CP-BC6, DYNEO™ DD-200F, DYNEO™ DD-201F, DYNEO™ DD-300F, DYNEO™ DD-BC4, DYNEO™ DD-BC6
4845.00 SEK
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
15430622
View Documents
JULABO™
9970242
CORIO™ CD-BC12, CORIO™ CP-1000F, CORIO™ CP-600F, CORIO™ CP-601F, CORIO™ CP-BC12, DYNEO™ DD-1000F, DYNEO™ DD-600F, DYNEO™ DD-601F, DYNEO™ DD-BC12
5705.00 SEK
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
Specifications
With built-in heat exchanger
Bath Cover