Learn More
Witeg™ Heavy Wall Centrifuge Tube
Centrifuge with confidence in these heavy wall tubes. Witeg™ Heavy Wall Centrifuge Tube is graduated and includes a screw cap top.
Brand:  Witeg™ 4 945 404
Additional Details : Weight : 0.13400kg
 View more versions of this product
Product Code. 15584279
  • 600.00 SEK / Each
Estimated Shipment: 27-08-2024
to see stock.
Add to basket
Description
Description
  • Heavy wall.
  • Graduated.
  • Round bottom
  • Screw cap closure.
Specifications
Specifications
100 mL (85 mL nominal)
Round
44 mm
98 mm
Screw Cap