Learn More
Witeg™ Heavy Wall Centrifuge Tube
Centrifuge with confidence in these heavy wall tubes. Witeg™ Heavy Wall Centrifuge Tube is graduated and includes a screw cap top.
Brand: Witeg™ 4 945 404
Additional Details : Weight : 0.13400kg
Description
- Heavy wall.
- Graduated.
- Round bottom
- Screw cap closure.
Specifications
100 mL (85 mL nominal) | |
Round | |
44 mm |
98 mm | |
Screw Cap |