missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hegsted Salt Mixture

Hegsted Salt Mixture is used for rodent diet.

1450.00 SEK - 26350.00 SEK

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11455642
View Documents
MP Biomedicals™
0290284002
2 kg
1450.00 SEK
2kg
Please to purchase this item. Need a web account? Register with us today!
11465642
View Documents
MP Biomedicals™
0290284010
10 kg
5505.00 SEK
10kg
Please to purchase this item. Need a web account? Register with us today!
11475642
View Documents
MP Biomedicals™
0290284050
50 kg
26350.00 SEK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Mineral Mix
  • For rodent diet
  • Powder
Specifications

Specifications

Animal Diet
Rodent diet
Videos
SDS
Documents

Documents

Product Certifications