Learn More
Heidolph™ Hei-FLOW Platinum Package
This package includes Hei-FLOW Precision 01, pump head SP quick 1.6, 1 m each Tygon and silicone tubing.
Brand: Heidolph™ 523-52019-00
Additional Details : Weight : 0.80000kg
Description
This package includes:
- Hei-FLOW Precision 01
- Pump head SP quick 1.6
- 1 m each Tygon™ (standard) and Silicone tubing (inner Ø 0.8 mm)
Specifications
EU Plug | |
LED | |
230 V |
Digital Variable Speed | |
IP33 |