Learn More
Heidolph™ Hei-TORQUE Silver Package
Package includes Hei-TORQUE Value, telescope stand, and clamp.
Brand: Heidolph™ 501-61019-00
23068.40 SEK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hei-TORQUE Value 100
- Telescope stand
- Clamp
Specifications
230 V | |
EU |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% - Multiple Manufacturers
Product Content Correction
Product Title
Heidolph™ Hei-TORQUE Silver Package > EU Plug
Spot an opportunity for improvement?Share a Content Correction