Learn More
Heidolph™ Instruments Hei-VAP Automatic Evaporation Package, EU-plug
The Hei-VAP Automatic Evaporation Package features a motor lift, condenser, Woulff bottle, and tubing set.
Brand:  Heidolph™ Instruments 574-37300-00
Additional Details : Weight : 20.30000kg
Product Code. 17313340
  • 124470.00 SEK / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
  • Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Vario Tec
  • Condenser for Rotavac Vario
  • Tubing set
  • Woulff bottle
  • AUTOaccurate-sensor
Specifications
Specifications
Motor Lift Rotary Evaporator
EU-plug