missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Instruments Hei-VAP Powder Drying-Chill Package, EU-plug

The Hei-VAP Powder Drying-Chill Package features a motor lift, chiller, powder flask, and tubing set.

Brand:  Heidolph™ Instruments 574-33300-00

Product Code. 17303340

  • 161675.00 SEK / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Valve Control
  • Vacuum valve
  • Chiller Hei-CHILL 600
  • RS 232 cable
  • Tubing set
  • Powder flask 2 L
  • Vapor tube with powder filter
  • Thermal fluid Kyro 30 (10 L)
Specifications

Specifications

Motor Lift Rotary Evaporator
EU-plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.