Learn More
Heidolph™ Instruments Hei-VAP Ultimate ML G3, EU-plug
The Hei-VAP Ultimate ML G3 allows for precise setting of rotation speed and heating bath temperature on the digital touch display or directly via the knobs on the operating panel.
Brand:  Heidolph™ Instruments 576-01300-00
Product Code. 17343360
  • 59280.00 SEK / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Precise setting of rotation speed and heating bath temperature on the digital touch display or – for quick access during an active process – directly via the knobs on the operating panel.
  • Central operating panel with 7 in. touch display
  • Motor lift model with G3 vertical glassware
  • Clear display of all values (Basic and Advanced modes)
  • With timer, limitation of max. temperature, multilingual menu and text display for error messages
  • Separate knobs for rotation and heating with locking function to prevent accidental readjusting
  • LED ring lights and pictogram to display the activity even from a distance
  • Integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller
  • USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
  • Optional display of vapor temperature
  • Subsequent upgrade to Hei-VAP Ultimate Control possible, including vacuum and chiller control
Specifications
Specifications
5 L
Motor Lift Rotary Evaporator
EU-plug