missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1336, Novus Biologicals™
Mouse Monoclonal Antibody
2450.00 SEK
Specifications
Antigen | Helicobacter Pylori |
---|---|
Clone | HP/1336 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
Applications | Flow Cytometry, ELISA, Immunofluorescence |
Description
Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Helicobacter Pylori | |
0.2 mg/ml | |
Flow Cytometry, ELISA, Immunofluorescence | |
Unconjugated | |
Mouse | |
Bacteria | |
H. pylori | |
IgG1 κ | |
Protein A or G purified |
HP/1336 | |
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Total sonicate of Helicobacter pylori | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title