missing translation for 'onlineSavingsMsg'
Learn More

Hematein, Thermo Scientific Chemicals

Product Code. 10222862
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475-25-2
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.27
MDL Number MFCD00036187
InChI Key HLUCICHZHWJHLL-UHFFFAOYNA-N
Synonym hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein
PubChem CID 10138
IUPAC Name 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one
SMILES OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O

Specifications

Melting Point 180°C
Color Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Beilstein 18,IV,3343
Merck Index 15,4669
Solubility Information Solubility in water: very slightly soluble. Other solubilities: slightly solubel in ethanol,ether,soluble in alkaline solutions,insoluble in benzene,chloroform
Formula Weight 300.26
Physical Form Crystalline Powder
Chemical Name or Material Hematein
Show More Show Less
Product Identifier
  • Hematein

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'