missing translation for 'onlineSavingsMsg'
Learn More
Heptyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals Product Code.: 15407989

Heptyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Product Code. 15407989
1 g
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy

Product Code. 15407989

Brand: Thermo Scientific Alfa Aesar J66576.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78617-12-6
Molecular Formula C13H26O6
Molecular Weight (g/mol) 278.345
MDL Number MFCD00063299
InChI Key NIDYWHLDTIVRJT-UJPOAAIJSA-N
Synonym heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
PubChem CID 448173
IUPAC Name (2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 1 g
Beilstein 8806
Formula Weight 278.34
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Heptyl beta-D-glucopyranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heptyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.