Learn More
Cole Parmer HF ACID EATER SPILL KIT
Brand:  Cole Parmer 86295-04
HF ACID EATER SPILL KIT
Product Code. 15252390
  • 5060.00 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • HF Acid Eater Liquid Neutralizer
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : calcium hydroxide, thymol blue