missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | HIP1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18370707
|
Bio-Techne
NBP3-05022-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18332514
|
Bio-Techne
NBP3-05022-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIP1 Polyclonal specifically detects HIP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HIP1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 410-690 of human HIP1 (NP_005329.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
3092 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title