missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Capto™ Phenyl (High Sub) Prepacked HIC Column
Hydrophobic interaction chromatography (HIC) media used in the capture and intermediate stages of protein purification, used for method optimization and parameter screening
Brand: Cytiva 28-9924-72
This item is not returnable.
View return policy
Description
- Highly cross-linked agarose matrix with average bead size of 75μm
- Has good flow properties and high loading capacities
- Bed height of 10cm designed to allow method optimization and parameter screening
- Connect easily in series to achieve 20cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Capto Phenyl (high sub) media is used for initial and intermediate step purifications requiring a chromatography medium with medium to high hydrophobicity
- Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Bed volume: 4.7mL
- Bed dimension: 7.7 × 100mm
- Flow rate: 1.2 to 2.7mL/min.
- Flow velocity: <600cm/hr.
- Average particle size: 75μm
- Degree of substitution: 27μmol/mL gel
- pH stability cleaning: 2 to 14
- Pressure/Flow specification: 300kPa at 600cm/hr., 1m diameter column, 20cm bed height
- Medium: BioProcess
- pH stability working range: 3 to 13
- Ligand: phenyl
- Binding capacity/mL chromatography medium: 27mg BSA /mL (at QB10%)
- Bed support: filter
- Material (column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
Specifications
Aqueous Buffer | |
4.7mL | |
7.7mm | |
3.93 in. | |
<600cm/hr. | |
Phenyl (∼27μM/mL drained medium) | |
3 to 13 | |
Highly Cross-linked Agarose | |
HIC Column |
HiScreen Capto Phenyl (high sub) column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
43.5psi | |
75μm | |
4° to 30°C | |
Optimization and parameter screening |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen Capto Phenyl (high sub), 1 x 4.7 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction