missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Capto™ PlasmidSelect
For purification of supercoiled DNA to high quality, for gene therapy and DNA vaccine applications
Brand: Cytiva 29201790
This item is not returnable.
View return policy
Description
- Allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms
- High-flow purification of supercoiled DNA from research scale to cGMP production
- Flexible process design due to a large operational window of flow velocities and bed heights
- Convenient small-scale purification, process development, and scale-up using prepacked HiScreen columns
- Capto™ PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production
- Higher throughput for improved productivity and process economy
- Hydrophilic properties of base matrix prevent non-specific binding
- BioProcess resin supported for industrial applications
Specifications
Gene Therapy and DNA Vaccine Applications | |
Prepacked Column | |
<2.3 mL/min (Max) | |
8 bar (0.8 MPa) | |
4.7 mL |
HiScreen | |
0.77 mm | |
10 cm | |
Capto™ PlasmidSelect |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.