missing translation for 'onlineSavingsMsg'
Learn More

Histamine diphosphate, MP Biomedicals™

Product Code. 11483020
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51-74-1
Molecular Formula C5H15N3O8P2
Molecular Weight (g/mol) 307.14
MDL Number MFCD00064316
InChI Key ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
Synonym histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate
PubChem CID 65513
ChEBI CHEBI:51193
IUPAC Name 2-(1H-imidazol-5-yl)ethan-1-amine; bis(phosphoric acid)
SMILES OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1

Specifications

Color White
Quantity 1 g
Formula Weight 325.15
Physical Form Crystalline Powder
Chemical Name or Material Histamine diphosphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'