missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-E Mouse anti-Human, Clone: 1A4G3, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HLA-E |
---|---|
Clone | 1A4G3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828495
|
Proteintech
66530-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16818495
|
Proteintech
66530-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody MEM-E/08 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results
The antibody MEM-E/08 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that antibody is remarkably specific for HLA-E, only with weak cross-reactivity with following classical MHC Class I molecules (MHC Class Ia): HLA-A24, -B7, -B27, -B51, -B54, -C7.Specifications
HLA-E | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EA1.2, EA2.1, HLA 6.2, HLA E, HLAE, HLA-E, MHC, MHC class I antigen E, QA1 | |
HLA-E | |
IgG2a | |
Protein A | |
Antibody |
1A4G3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P13747 | |
3133 | |
HLA-E Fusion Protein Ag6724 | |
Primary | |
-20°C | |
HLA-E |