missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP UL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561983
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Immunogen sequence: PTAQTYPQPS YNQYQQYAQQ WNQYYQNQGQ WPPYYGNYDY GSYSGNTQGG TSTQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells.This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.
Specifications
hnRNP UL1 | |
Polyclonal | |
Unconjugated | |
Hnrnpul1 | |
Adenovirus early region 1B-associated protein 5; E130317O14Rik; E1B 55kDa associated protein 5; E1B-55 kDa-associated protein 5; E1BAP5; E1B-AP5; EIB-55kDa associated protein 5; heterogeneous nuclear ribonucleoprotein U like 1; heterogeneous nuclear ribonucleoprotein U-like 1; heterogeneous nuclear ribonucleoprotein U-like protein 1; Hnrnpul; Hnrnpul1; HNRPUL1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein U-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUJ2 | |
Hnrnpul1 | |
Recombinant protein corresponding to Human hnRNP UL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
hnRNP UL1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Spot an opportunity for improvement?Share a Content Correction