Learn More
Hoefer™ Clamp Thumbscrews
Accessories for Vertical Protein Electrophoresis Systems.
Brand: Hoefer™ SE6003U-2
390.55 SEK valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- 12 Pk.
- For 8 cm and 16 cm clamp assemblies
Specifications
Vertical Gel Electrophoresis Unit | |
Clamp Thumbscrew |