Learn More
Hoefer™ Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
3270.00 SEK - 6940.00 SEK
Product Code | Brand | For Use With (Equipment) | Includes | No. of Gels | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | No. of Gels | Price | Quantity & Availability | |||
12039857
|
Hoefer™
SE235 |
SE 260, MiniVE™ Electrophoresis System | Caster body with gasket, face plate with inlet port, 10 x 10.5cm rectangular Glass plates, 10 x 10.5cm notched Alumina plates, spring clamps, Wax Paper, space-saver plate, Polycarbonate filler sheets, black Vinyl cap, filler plug set | 1 to 4 |
5640.00 SEK
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
10772677
|
Hoefer™
SE235 |
SE 260, MiniVE™ Electrophoresis System | Caster body with gasket, face plate with inlet port, 10 x 10.5cm rectangular Glass plates, 10 x 10.5cm notched Alumina plates, spring clamps, Wax Paper, space-saver plate, Polycarbonate filler sheets, black Vinyl cap, filler plug set | 1 to 4 | N/A | ||||
12064837
|
Hoefer™
SE245 |
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System | Casting cradle, clamp assemblies, sealing gasket set, cams | 1 to 2 |
3270.00 SEK
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||
10565965
|
Hoefer™
SE245 |
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System | Casting cradle, clamp assemblies, sealing gasket set, cams | 1 to 2 | N/A | ||||
12099837
|
Hoefer™
SE215 |
SE 250 Electrophoresis System | Caster body with gasket, face plate with inlet port, 10 x 8cm rectangular Glass plates, 10 x 8cm notched Alumina plates, spring clamps, Wax Paper, space-saver plate, Polycarbonate filler sheets, black Vinyl cap, filler plug set | 4 to 10 |
4865.00 SEK
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
12059837
|
Hoefer™
SE275 |
SE 250 Mini Vertical Gel Electrophoresis Unit | Casting cradle with gasket, face, Glass, alumina, space saver plates, filler sheets, plugs | 1 to 4 |
5385.00 SEK
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||
Description
SE245 Dual-Gel Caster
- For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
- Suitable for single concentration and gradient gels
- SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
- SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
Specifications
Gel Caster |