missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Spacel™ 3000 RA/EBJ Coveralls with Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Product Type: Coveralls
- Material: Polyethylene
- Gender: Unisex
- Color: Yellow
- Disposable: Yes
- Sterility: Non-Sterile
- Resistance: Chemical, Abrasion, Tear, and Perforation
- Certification: EC Category 3, Protection Level: 3, 4, 5, 6b
- Hood, Wrist, and Ankle Type: Elasticated
- Closure Type: Sealed Zipper/Double Adhesive Flap
- Seam Type: Sealed
- For Use With: Chemistry, Petrochemistry, In-Shore/Off-Shore Oil Industry, Industrial Maintenance, Pharmaceutical Industry, Spoiled Beach Cleaning, Transportation of Hazardous Materials, Industrial Sub-Contracting Cleaning, Sanitation, Waste Processing, Emergency Services
Specifications
Specifications
Product Type | Hooded Coveralls |
Size | X-Large |
Gender | Unisex |
Material | Polyethylene |
Color | Yellow |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Hood Type | Elastic |
Closure Type | Sealed Zipper with Double-adhesive Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'