Learn More
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, D-100
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, D-100 is compact but designed with an ideal chamber capacity for most standard media bottles and Erlenmeyer flasks.
Brand: Systec™ 2100
This item is not returnable.
View return policy
Description
- Standard features:
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- Temperature and pressure range 140°C, 4 bar (except for Systec D-23)
- LCD display and fully automatic microprocessor control
- Code-secured access rights for changing parameters and further safety-relevant intervention
- Flexible PT-100 temperature sensor
- Optional: Integrated printer for batch documentation
Specifications
110 L (Total), 100 L (Nominal) | |
500 mm | |
Horizontal, Front-loading Benchtop Autoclave |
500 mm | |
D100 |