Learn More
Brady™ Hot Temperature Indicating Label
Temperature Indicating Labels are a cost-effective solution for temperature checks during maintenance or for trouble spotting during research and development.
Brand: Brady™ 195844
243.20 SEK valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low profile allows for application on components, on industrial and household applications such as electric pumps, rotating equipment, brakes and refrigerators, in cold chains and hot spaces or in areas where measuring temperature becomes difficult or impractical
Specifications
TIL-12-50C-DIA | |
Black, Red | |
37.08 mm |
Permanent Acrylic | |
Polyester (B-7518) |