missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 Mouse anti-Virus, Clone: HPV16/1295, Novus Biologicals™
Mouse Monoclonal Antibody
2450.00 SEK
Specifications
Antigen | HPV16 |
---|---|
Clone | HPV16/1295 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
HPV16 Monoclonal specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry-Paraffin.Specifications
HPV16 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Virus | |
Human Papilloma Virus Type 16 | |
IgG1 κ | |
Protein A or G purified |
HPV16/1295 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
HPV-16 E6 peptide (MHQKRTAMFQDPPQERPRKLPQLC) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title