missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | HRAS |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323614
|
Bio-Techne
NBP3-04486-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18363734
|
Bio-Techne
NBP3-04486-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HRAS Polyclonal specifically detects HRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HRAS | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
3265 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Metastasis, mTOR Pathway | |
PBS with 50% glycerol, pH7.3. | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human HRAS (NP_005334.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title