Learn More
GeneBLAzer™ HRH3-bla U2OS DA Assay Kit
GeneBLAzer™ HRH3-bla U2OS DA Assay Kit contains the human Histamine Receptor H3 (HRH3) linked to a TEV protease site and a Gal4-VP16 transcription factor.
Brand: GeneBLAzer™ K1509
Additional Details : Weight : 0.05000kg
Description
- Gene Symbol: HRH3
- Accession Number: NM_007232
- Tail from AVPR2 GPCR
- Population Status: Pool of Stable Cells
- This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
- The Tango™ HRH3-bla U2OS cells and the Tango™ HRH3-bla U2OS DA cells have been functionally validated for Z' factor and EC50 concentrations of Methylhistamine.
- In addition, Tango™ HRH3-bla U2OS cells have been tested for assay performance under variable conditions.
- These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
- DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
Specifications
Should be stored in liquid nitrogen immediately upon receipt | |
GPCRs | |
1 Plate |
Fluorescent | |
HRH3-bla U2OS DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70 μg, Solution D, Tango™ HRH3-bla U2OS Assay Protocol, LiveBLAzer™FRET B/G Loading Protocol, Certificate of Analysis | |
Division-Arrested Cells Assay Kit |