Learn More
Julabo™ HST Booster Heater for Ultra-Low Refrigerated Circulators
Brand: Julabo™ 8810012
This item is not returnable.
View return policy
Specifications
FP52-SL, FP52-SL, FP52-SL-150C, FP55-SL, FP55-SL-150C Ultra-Low Refrigerated-Heating Circulator | |
HST Booster Heater | |
Ideal for controlling the temperature of large or distant objects |