missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-3 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
818.00 SEK - 30750.00 SEK
Specifications
Accession Number | P81534 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 414325 |
Molecular Weight (g/mol) | 5.1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17872793
|
Gibco™
300-52-100UG |
100 μg |
6150.00 SEK
100µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17882933
|
Gibco™
300-52-1MG |
1 mg |
30750.00 SEK
1mg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17842093
|
Gibco™
300-52-20UG |
20 μg |
2015.00 SEK
20µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17892693
|
Gibco™
300-52-250UG |
250 μg |
10770.00 SEK
250µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17832843
|
Gibco™
300-52-500UG |
500 μg |
18430.00 SEK
500µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17882193
|
Gibco™
300-52-50UG |
50 μg |
3705.00 SEK
50µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17842133
|
Gibco™
300-52-5UG |
5 μg |
818.00 SEK
5µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
P81534 | |
protein with no preservative | |
5.1 | |
E. coli | |
RUO | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
414325 | |
Human BD-3 | |
-20°C | |
<1 EU/μg | |
DEFB103A | |
Unconjugated | |
GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |